Assay Details
Target Gene Details
Entrez Gene ID: | 360200 |
Gene Name: | transmembrane protease, serine 9 |
Gene Aliases: |
- |
Location: |
Chr.19:2385254-2426261 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS9 | NM_182973.1 | 3 | 373 | NP_892018.1 |
| XM_011527978.2 | 5 | 736 | XP_011526280.1 | |
| AB109390.1 | 4 | 490 | ||
| AJ488946.1 | 3 | 373 | CAD35758.1 | |
| AJ488947.1 | 3 | 373 | CAD35759.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160562 | Chr.19:2328434 - 2460264 on Build GRCh38 | Deletion |
|
| nsv1064930 | Chr.19:2380093 - 2592252 on Build GRCh38 | Gain |
|
| esv3583208 | Chr.19:2389734 - 2409683 on Build GRCh38 | Loss |
|
| nsv953944 | Chr.19:2283702 - 2525802 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map