Assay Details
Target Gene Details
Entrez Gene ID: | 3425 |
Gene Name: | iduronidase, alpha-L- |
Gene Aliases: |
IDA, MPS1 |
Location: |
Chr.4:986997-1004557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDUA | NM_000203.4 | NP_000194.2 | ||
| NR_110313.1 | ||||
| XM_017008163.1 | XP_016863652.1 | |||
| AB208901.1 | 3 | 2463 | BAD92138.1 | |
| AK125223.1 | ||||
| AK291816.1 | ||||
| BC029959.1 | AAH29959.1 | |||
| DA057148.1 | ||||
| M74715.1 | AAA81589.1 |
Target Gene Details
Entrez Gene ID: | 10861 |
Gene Name: | solute carrier family 26 member 1 |
Gene Aliases: |
CAON, EDM4, SAT-1, SAT1 |
Location: |
Chr.4:979073-993440 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A1 | NM_022042.3 | NP_071325.2 | ||
| NM_134425.2 | NP_602297.1 | |||
| NM_213613.3 | NP_998778.1 | |||
| AF297659.2 | AAG22075.1 | |||
| AK292747.1 | ||||
| AY124771.1 | AAM94171.1 | |||
| BC015517.2 | AAH15517.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482086 | Chr.4:987659 - 993436 on Build GRCh38 | Gain |
|
| nsv1003219 | Chr.4:776106 - 1111676 on Build GRCh38 | Gain |
|
| nsv822428 | Chr.4:968896 - 1028144 on Build GRCh38 | Gain |
|
| nsv593226 | Chr.4:948218 - 1025223 on Build GRCh38 | Loss |
|
| esv3599384 | Chr.4:974637 - 1010875 on Build GRCh38 | Gain |
|
| nsv822425 | Chr.4:546316 - 1420011 on Build GRCh38 | Loss |
|
| nsv4196 | Chr.4:961755 - 1004599 on Build GRCh38 | Deletion |
|
| nsv950328 | Chr.4:988213 - 1101612 on Build GRCh38 | Deletion |
|
| nsv1078530 | Chr.4:987811 - 994112 on Build GRCh38 | Duplication |
|
| nsv469998 | Chr.4:815805 - 1093743 on Build GRCh38 | Loss |
|
| nsv1008352 | Chr.4:819605 - 1041628 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114936403] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map