Assay Details
Target Gene Details
Entrez Gene ID: | 7352 |
Gene Name: | uncoupling protein 3 |
Gene Aliases: |
SLC25A9 |
Location: |
Chr.11:74000281-74009237 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UCP3 | NM_003356.3 | 7 | 1663 | NP_003347.1 |
| BC008392.1 | 5 | 1202 | AAH08392.1 | |
| BU618868.1 | 1 | 693 |
More Information
Set Membership: |
|

Genomic Map