Assay Details
Target Gene Details
Entrez Gene ID: | 440435 |
Gene Name: | G protein-coupled receptor 179 |
Gene Aliases: |
CSNB1E, GPR158L, GPR158L1 |
Location: |
Chr.17:38325610-38343810 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR179 | NM_001004334.3 | 11 | 5922 | NP_001004334.3 |
| AY584611.1 | 11 | 5922 | AAS91740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428341 | Chr.17:37855347 - 38600766 on Build GRCh38 | Gain+Loss |
|
| esv2758688 | Chr.17:37706608 - 38600766 on Build GRCh38 | Gain+Loss |
|
| esv33106 | Chr.17:38299424 - 38357921 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map