Assay Details
Target Gene Details
Entrez Gene ID: | 1508 |
Gene Name: | cathepsin B |
Gene Aliases: |
APPS, CPSB |
Location: |
Chr.8:11842524-11868150 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTSB | NM_001317237.1 | 10 | 2171 | NP_001304166.1 |
| NM_001908.4 | 10 | 2202 | NP_001899.1 | |
| NM_147780.3 | 12 | 2364 | NP_680090.1 | |
| NM_147781.3 | 11 | 2321 | NP_680091.1 | |
| NM_147782.3 | 11 | 2290 | NP_680092.1 | |
| NM_147783.3 | 11 | 2276 | NP_680093.1 | |
| XM_006716244.2 | 12 | 2388 | XP_006716307.1 | |
| XM_006716245.2 | 10 | 2492 | XP_006716308.1 | |
| XM_011543812.2 | 12 | 2297 | XP_011542114.1 | |
| XM_017013097.1 | 11 | 2278 | XP_016868586.1 | |
| XM_017013098.1 | 11 | 2278 | XP_016868587.1 | |
| XM_017013099.1 | 12 | 2320 | XP_016868588.1 | |
| XM_017013100.1 | 11 | 2222 | XP_016868589.1 | |
| XM_017013101.1 | 7 | 1745 | XP_016868590.1 | |
| AK097384.1 | 5 | 2132 | ||
| AK098296.1 | 1 | 163 | ||
| AK128472.1 | 1 | 2795 | ||
| BC095408.1 | 10 | 2139 | AAH95408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034132 | Chr.8:11799524 - 12340733 on Build GRCh38 | Gain |
|
| nsv610293 | Chr.8:11841238 - 12072225 on Build GRCh38 | Gain |
|
| esv34042 | Chr.8:11840828 - 11869308 on Build GRCh38 | Loss |
|
| nsv1018042 | Chr.8:11809987 - 12024876 on Build GRCh38 | Gain |
|
| nsv1032739 | Chr.8:11786294 - 11858964 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map