Assay Details
Target Gene Details
Entrez Gene ID: | 4882 |
Gene Name: | natriuretic peptide receptor 2 |
Gene Aliases: |
AMDM, ANPRB, ANPb, ECDM, GUC2B, GUCY2B, NPRB, NPRBi, SNSK |
Location: |
Chr.9:35752945-35809731 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 26 - Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPR2 | NM_003995.3 | NP_003986.2 | ||
| XM_005251478.3 | XP_005251535.1 | |||
| XM_005251479.4 | XP_005251536.1 | |||
| XM_011517890.2 | XP_011516192.1 | |||
| XM_011517891.2 | XP_011516193.1 | |||
| XM_011517892.2 | XP_011516194.1 | |||
| XM_011517893.2 | XP_011516195.1 | |||
| XM_011517895.2 | XP_011516197.1 | |||
| XM_017014745.1 | XP_016870234.1 | |||
| XM_017014746.1 | XP_016870235.1 | |||
| XM_017014747.1 | XP_016870236.1 | |||
| XM_017014748.1 | XP_016870237.1 | |||
| XM_017014749.1 | XP_016870238.1 | |||
| XM_017014750.1 | XP_016870239.1 | |||
| AJ005282.1 | CAA06466.1 | |||
| BC023017.1 | AAH23017.1 | |||
| BQ446138.1 | ||||
| L13436.1 |
Target Gene Details
Entrez Gene ID: | 26206 |
Gene Name: | sperm associated antigen 8 |
Gene Aliases: |
BS-84, CILD28, CT142, HSD-1, SMP1, SPAG3, hSMP-1 |
Location: |
Chr.9:35807785-35812262 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPAG8 | NM_172312.1 | NP_758516.1 | ||
| BC019247.1 | AAH19247.1 | |||
| S83157.1 | AAB46833.1 | |||
| U12978.2 | AAA21129.2 |
More Information
Set Membership: |
|

Genomic Map