Assay Details
Target Gene Details
Entrez Gene ID: | 1659 |
Gene Name: | DEAH-box helicase 8 |
Gene Aliases: |
DDX8, HRH1, PRP22, PRPF22 |
Location: |
Chr.17:43483865-43544787 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX8 | NM_001302623.1 | 1 | 97 | NP_001289552.1 |
| NM_001322216.1 | 1 | 149 | NP_001309145.1 | |
| NM_001322217.1 | 1 | 149 | NP_001309146.1 | |
| NM_001322218.1 | 1 | 149 | NP_001309147.1 | |
| NM_001322219.1 | 1 | 149 | NP_001309148.1 | |
| NM_001322220.1 | 1 | 149 | NP_001309149.1 | |
| NM_001322221.1 | 1 | 149 | NP_001309150.1 | |
| NM_004941.2 | 1 | 97 | NP_004932.1 | |
| NR_136225.1 | 1 | 149 | ||
| NR_136226.1 | 1 | 149 | ||
| NR_136227.1 | 1 | 149 | ||
| NR_136228.1 | 1 | 149 | ||
| AK291664.1 | ||||
| AK294046.1 | 1 | 149 | ||
| AK303319.1 | 1 | 51 | ||
| BC020697.1 | 1 | 54 | ||
| BC044586.1 | ||||
| BC047327.1 | ||||
| D50487.1 | 1 | 49 | BAA09078.1 | |
| DC310785.1 | 1 | 149 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761959 | Chr.17:43322294 - 43565935 on Build GRCh38 | Gain |
|
| nsv833453 | Chr.17:43431629 - 43612561 on Build GRCh38 | Gain+Loss |
|
| nsv470588 | Chr.17:43346885 - 43616240 on Build GRCh38 | Gain |
|
| esv3640625 | Chr.17:43361371 - 43577834 on Build GRCh38 | Gain |
|
| nsv833451 | Chr.17:43329618 - 43499146 on Build GRCh38 | Loss |
|
| nsv1065802 | Chr.17:43338154 - 43584333 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map