Assay Details
Target Gene Details
Entrez Gene ID: | 1043 |
Gene Name: | CD52 molecule |
Gene Aliases: |
CDW52 |
Location: |
Chr.1:26317920-26320523 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD52 | NM_001803.2 | NP_001794.2 | ||
| A23013.1 | CAA01644.1 | |||
| BC000644.2 | AAH00644.1 | |||
| BI835519.1 | ||||
| BT007116.1 | AAP35780.1 | |||
| CR456874.1 | CAG33155.1 | |||
| FJ460513.1 | ||||
| KP176645.1 | ||||
| X62466.1 | CAA44323.1 | |||
| X67699.1 | CAA47929.1 |
More Information
Set Membership: |
|

Genomic Map