Assay Details
Target Gene Details
Entrez Gene ID: | 323 |
Gene Name: | amyloid beta precursor protein binding family B member 2 |
Gene Aliases: |
FE65L, FE65L1 |
Location: |
Chr.4:40810027-41216714 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv10486 | Chr.4:40747595 - 40935088 on Build GRCh38 | Gain |
|
| nsv4307 | Chr.4:40796057 - 40823222 on Build GRCh38 | Deletion |
|
| esv3348981 | Chr.4:40814778 - 40816576 on Build GRCh38 | Duplication |
|
| nsv829912 | Chr.4:40720092 - 40840600 on Build GRCh38 | Loss |
|
| nsv999749 | Chr.4:40768481 - 40820050 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs554579308] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map