Assay Details
Target Gene Details
Entrez Gene ID: | 29095 |
Gene Name: | ORMDL sphingolipid biosynthesis regulator 2 |
Gene Aliases: |
HSPC160, MST095, MSTP095, adoplin-2 |
Location: |
Chr.12:55818022-55821175 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORMDL2 | NM_014182.4 | NP_054901.1 | ||
| AB064961.1 | BAC11712.1 | |||
| AF161509.1 | AAF29124.1 | |||
| AF173375.1 | AAQ13618.1 | |||
| AF395705.1 | AAM43504.1 | |||
| AF395706.1 | AAM43505.1 | |||
| AF395707.1 | AAM43506.1 | |||
| AK223180.1 | BAD96900.1 | |||
| AK314046.1 | ||||
| BC012543.2 | AAH12543.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832423 | Chr.12:55687038 - 55859914 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map