Assay Details
Target Gene Details
Entrez Gene ID: | 57154 |
Gene Name: | SMAD specific E3 ubiquitin protein ligase 1 |
Gene Aliases: |
- |
Location: |
Chr.7:99027435-99144120 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMURF1 | NM_001199847.1 | NP_001186776.1 | ||
| NM_020429.2 | NP_065162.1 | |||
| NM_181349.2 | NP_851994.1 | |||
| XM_017012457.1 | XP_016867946.1 | |||
| AB046845.1 | ||||
| AF199364.2 | AAF08298.2 | |||
| AF464850.1 | AAM90910.1 | |||
| AL834242.1 | CAD38919.1 | |||
| BC136804.1 | ||||
| BC144414.1 | ||||
| BC152468.1 |
More Information
Set Membership: |
|

Genomic Map