Assay Details
Target Gene Details
Entrez Gene ID: | 222826 |
Gene Name: | family with sequence similarity 217 member A |
Gene Aliases: |
C6orf146 |
Location: |
Chr.6:4068352-4087344 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM217A | NM_173563.2 | NP_775834.2 | ||
| XM_005248951.3 | XP_005249008.1 | |||
| XM_006715025.2 | XP_006715088.1 | |||
| XM_006715026.3 | XP_006715089.1 | |||
| XM_011514413.2 | XP_011512715.1 | |||
| XM_011514414.2 | XP_011512716.1 | |||
| XM_011514415.1 | XP_011512717.1 | |||
| XM_011514416.2 | XP_011512718.1 | |||
| XM_011514417.1 | XP_011512719.1 | |||
| XM_017010480.1 | XP_016865969.1 | |||
| XM_017010481.1 | XP_016865970.1 | |||
| XM_017010482.1 | XP_016865971.1 | |||
| AK097426.1 | ||||
| BC039349.1 | AAH39349.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600860 | Chr.6:4052149 - 4096444 on Build GRCh38 | Loss |
|

Genomic Map