Assay Details
Target Gene Details
Entrez Gene ID: | 80344 |
Gene Name: | DDB1 and CUL4 associated factor 11 |
Gene Aliases: |
GL014, PRO2389, WDR23 |
Location: |
Chr.14:24114697-24125242 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF11 | NM_001163484.1 | 1 | 169 | NP_001156956.1 |
| NM_025230.4 | 1 | 89 | NP_079506.3 | |
| NR_028099.1 | 1 | 89 | ||
| AB209073.1 | 1 | 75 | BAD92310.1 | |
| AK074296.1 | BAB85041.1 | |||
| AK307849.1 | ||||
| BC067132.1 | AAH67132.1 | |||
| BP245093.1 | 1 | 89 | ||
| CD644652.1 | 2 | 319 |
Target Gene Details
Entrez Gene ID: | 4901 |
Gene Name: | neural retina leucine zipper |
Gene Aliases: |
D14S46E, NRL-MAF, RP27 |
Location: |
Chr.14:24078693-24114924 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NRL | XM_005267708.4 | 1 | 60 | XP_005267765.1 |
| XM_005267710.3 | XP_005267767.1 | |||
| XM_017021350.1 | 1 | 60 | XP_016876839.1 | |
| BX161381.1 | 1 | 150 | CAD61873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564057 | Chr.14:24056425 - 24149650 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map