Assay Details
Target Gene Details
Entrez Gene ID: | 10817 |
Gene Name: | fibroblast growth factor receptor substrate 3 |
Gene Aliases: |
FRS2-beta, FRS2B, FRS2beta, SNT-2, SNT2 |
Location: |
Chr.6:41770176-41779905 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRS3 | NM_006653.4 | 7 | 1703 | NP_006644.1 |
| XM_011514254.2 | 7 | 2556 | XP_011512556.1 | |
| XM_011514255.2 | 7 | 2551 | XP_011512557.1 | |
| XM_017010193.1 | 7 | 3311 | XP_016865682.1 | |
| AF036718.1 | 6 | 1582 | AAB92555.1 | |
| BC010611.1 | 6 | 1521 | AAH10611.1 | |
| CR457026.1 | CAG33307.1 |
More Information
Set Membership: |
|

Genomic Map