Assay Details
Target Gene Details
Entrez Gene ID: | 26580 |
Gene Name: | BSCL2, seipin lipid droplet biogenesis associated |
Gene Aliases: |
GNG3LG, HMN5, PELD, SPG17 |
Location: |
Chr.11:62690262-62709619 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BSCL2 | NM_001122955.3 | NP_001116427.1 | ||
| NM_001130702.2 | NP_001124174.2 | |||
| NM_032667.6 | NP_116056.3 | |||
| NR_037948.1 | ||||
| NR_037949.1 | ||||
| AF052149.1 | ||||
| AK027524.1 | BAB55175.1 | |||
| AK075317.1 | BAC11543.1 | |||
| AK223606.1 | BAD97326.1 | |||
| AK225029.1 | ||||
| AK315423.1 | ||||
| BC004911.2 | AAH04911.1 | |||
| BC012140.1 | AAH12140.1 | |||
| BC041640.1 | AAH41640.1 | |||
| BC093048.1 | AAH93048.1 | |||
| CD107822.1 | ||||
| DB296305.1 | ||||
| KU178595.1 | ||||
| KU178596.1 |
Target Gene Details
Entrez Gene ID: | 100534595 |
Gene Name: | HNRNPUL2-BSCL2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:62690262-62727384 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HNRNPUL2-BSCL2 | NR_037946.1 | |||
| AK122942.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss |
|
| nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map