Assay Details
Target Gene Details
Entrez Gene ID: | 7760 |
Gene Name: | zinc finger protein 213 |
Gene Aliases: |
CR53, ZKSCAN21, ZSCAN53 |
Location: |
Chr.16:3135056-3142804 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF213 | NM_001134655.1 | NP_001128127.1 | ||
| NM_004220.2 | NP_004211.1 | |||
| NR_104432.1 | ||||
| XM_011522652.2 | XP_011520954.1 | |||
| AF017433.1 | AAB70531.1 | |||
| AK055337.1 | ||||
| AK289834.1 | ||||
| AK291789.1 | ||||
| AK297454.1 | ||||
| BC007287.1 | AAH07287.1 | |||
| KU178402.1 | ||||
| KU178403.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509589 | Chr.16:3045935 - 3169200 on Build GRCh38 | Insertion |
|
| nsv571249 | Chr.16:3115424 - 3251897 on Build GRCh38 | Gain |
|
| nsv571248 | Chr.16:3034108 - 3165103 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map