Assay Details
Target Gene Details
Entrez Gene ID: | 221692 |
Gene Name: | phosphatase and actin regulator 1 |
Gene Aliases: |
RPEL, RPEL1, dJ257A7.2 |
Location: |
Chr.6:12716635-13290484 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHACTR1 | NM_001242648.2 | NP_001229577.1 | ||
| NM_001322308.1 | NP_001309237.1 | |||
| NM_030948.3 | 2 | 334 | NP_112210.1 | |
| XM_005248934.2 | 2 | 176 | XP_005248991.1 | |
| XM_017010454.1 | 2 | 176 | XP_016865943.1 | |
| XM_017010455.1 | XP_016865944.1 | |||
| XM_017010457.1 | XP_016865946.1 | |||
| XM_017010459.1 | 2 | 176 | XP_016865948.1 | |
| XM_017010460.1 | XP_016865949.1 | |||
| XM_017010461.1 | XP_016865950.1 | |||
| AK090769.1 | ||||
| AK290017.1 | ||||
| AK295263.1 | ||||
| DA253448.1 | 2 | 176 | ||
| DA330657.1 |
More Information
Set Membership: |
|

Genomic Map