Assay Details
Target Gene Details
Entrez Gene ID: | 8665 |
Gene Name: | eukaryotic translation initiation factor 3 subunit F |
Gene Aliases: |
EIF3S5, eIF3-p47 |
Location: |
Chr.11:7987320-7996171 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3F | NM_003754.2 | NP_003745.1 | ||
| AK093511.1 | 3 | 610 | ||
| AK095574.1 | BAC04577.1 | |||
| AK289637.1 | ||||
| AK291354.1 | ||||
| AK293826.1 | ||||
| AK297621.1 | ||||
| BT006894.1 | AAP35540.1 | |||
| CB141463.1 | ||||
| CD050033.1 | ||||
| CR456959.1 | CAG33240.1 | |||
| U94855.1 | AAD03467.1 |
More Information
Set Membership: |
|

Genomic Map