Assay Details
Target Gene Details
Entrez Gene ID: | 57722 |
Gene Name: | immunoglobulin superfamily DCC subclass member 4 |
Gene Aliases: |
DDM36, NOPE |
Location: |
Chr.15:65381480-65423072 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGDCC4 | NM_020962.2 | 20 | 4440 | NP_066013.1 |
| XM_011521845.2 | 20 | 4620 | XP_011520147.1 | |
| XM_011521846.2 | 20 | 4247 | XP_011520148.1 | |
| XM_011521847.1 | 20 | 4307 | XP_011520149.1 | |
| XM_017022448.1 | 20 | 4625 | XP_016877937.1 | |
| AB046848.1 | 14 | 3500 | BAB13454.1 | |
| AB052622.1 | 20 | 4440 | BAB86306.1 | |
| AK124045.1 | 3 | 1363 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036360 | Chr.15:65333881 - 65390204 on Build GRCh38 | Gain |
|

Genomic Map