Assay Details
Target Gene Details
Entrez Gene ID: | 57409 |
Gene Name: | MIF4G domain containing |
Gene Aliases: |
AD023, MIFD, SLIP1 |
Location: |
Chr.17:75266228-75271292 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIF4GD | NM_001242498.1 | 7 | 1074 | NP_001229427.1 |
| NM_001242500.1 | 6 | 976 | NP_001229429.1 | |
| NM_001242501.1 | 5 | 874 | NP_001229430.1 | |
| NM_020679.3 | 7 | 1053 | NP_065730.2 | |
| XM_005257530.1 | 7 | 1195 | XP_005257587.1 | |
| XM_005257532.4 | 6 | 951 | XP_005257589.1 | |
| XM_005257533.1 | 6 | 1050 | XP_005257590.1 | |
| XM_011525051.2 | 7 | 1290 | XP_011523353.1 | |
| XM_011525052.2 | 7 | 1269 | XP_011523354.1 | |
| XM_011525053.2 | 6 | 1167 | XP_011523355.1 | |
| XM_011525054.2 | 6 | 1134 | XP_011523356.1 | |
| XM_011525055.2 | 5 | 1011 | XP_011523357.1 | |
| XM_011525057.2 | 4 | 714 | XP_011523359.1 | |
| XM_017024888.1 | 6 | 1860 | XP_016880377.1 | |
| XM_017024889.1 | 6 | 1644 | XP_016880378.1 | |
| XM_017024890.1 | 6 | 1670 | XP_016880379.1 | |
| XM_017024891.1 | 6 | 1582 | XP_016880380.1 | |
| XM_017024893.1 | 6 | 1446 | XP_016880382.1 | |
| XM_017024894.1 | 6 | 1081 | XP_016880383.1 | |
| XM_017024895.1 | 6 | 897 | XP_016880384.1 | |
| AF225422.1 | 5 | 1071 | AAG09724.1 | |
| BC033759.1 | 6 | 952 | AAH33759.1 | |
| BU190758.1 | ||||
| BX404175.2 | 7 | 996 | ||
| CA488406.1 | 5 | 874 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map