Assay Details
Target Gene Details
Entrez Gene ID: | 80830 |
Gene Name: | apolipoprotein L6 |
Gene Aliases: |
APOL-VI, APOLVI |
Location: |
Chr.22:35648317-35668409 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOL6 | NM_030641.3 | 3 | 9495 | NP_085144.1 |
| XM_011530392.2 | 4 | 9672 | XP_011528694.1 | |
| AK097266.1 | 1 | 1607 | ||
| BM982940.1 |
More Information
Set Membership: |
|

Genomic Map