Assay Details
Target Gene Details
Entrez Gene ID: | 55696 |
Gene Name: | RNA binding motif protein 22 |
Gene Aliases: |
Cwc2, ZC3H16, fSAP47 |
Location: |
Chr.5:150690790-150701107 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBM22 | NM_018047.2 | NP_060517.1 | ||
| AK001152.1 | BAA91521.1 | |||
| AK297019.1 | ||||
| AL136933.1 | CAB66867.1 | |||
| BC003402.1 | AAH03402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
|
| esv29484 | Chr.5:150679152 - 150700437 on Build GRCh38 | Loss |
|
| nsv428129 | Chr.5:150641519 - 150906180 on Build GRCh38 | Loss |
|
| esv33898 | Chr.5:150657871 - 150766463 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map