Assay Details
Target Gene Details
Entrez Gene ID: | 27173 |
Gene Name: | solute carrier family 39 member 1 |
Gene Aliases: |
ZIP1, ZIRTL |
Location: |
Chr.1:153959099-153968184 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC39A1 | NM_001271957.1 | 4 | 880 | NP_001258886.1 |
| NM_001271958.1 | 4 | 717 | NP_001258887.1 | |
| NM_001271959.1 | 4 | 886 | NP_001258888.1 | |
| NM_001271960.1 | 5 | 778 | NP_001258889.1 | |
| NM_001271961.1 | 3 | 586 | NP_001258890.1 | |
| NM_014437.4 | 5 | 1024 | NP_055252.2 | |
| AF132942.1 | 3 | 574 | AAD27717.1 | |
| AF151829.1 | 4 | 595 | AAD34066.1 | |
| AJ243649.1 | 4 | 611 | CAB59979.1 | |
| AJ271671.1 | 4 | 625 | CAB82784.1 | |
| AK074943.1 | 4 | 834 | ||
| AK075257.1 | 4 | 648 | BAC11502.1 | |
| AK293389.1 | 3 | 494 | ||
| AK297983.1 | 5 | 592 | ||
| AK300244.1 | 5 | 584 | ||
| BC002563.2 | 4 | 599 | AAH02563.1 | |
| BC003152.2 | 3 | 571 | AAH03152.1 | |
| BC007886.2 | 4 | 596 | AAH07886.1 | |
| BC014303.2 | 5 | 1004 | AAH14303.1 | |
| CR457217.1 | 3 | 538 | CAG33498.1 |
More Information
Set Membership: |
|

Genomic Map