Assay Details
Target Gene Details
Entrez Gene ID: | 7994 |
Gene Name: | lysine acetyltransferase 6A |
Gene Aliases: |
MOZ, MRD32, MYST-3, MYST3, RUNXBP2, ZC2HC6A, ZNF220 |
Location: |
Chr.8:41929479-42052026 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT6A | NM_006766.4 | 17 | 7650 | NP_006757.2 |
| XM_011544656.2 | 19 | 7876 | XP_011542958.1 | |
| XM_011544657.2 | 18 | 7743 | XP_011542959.1 | |
| XM_011544658.2 | 19 | 7832 | XP_011542960.1 | |
| XM_011544659.2 | 19 | 7855 | XP_011542961.1 | |
| XM_017013863.1 | 18 | 7744 | XP_016869352.1 | |
| XM_017013864.1 | 18 | 7700 | XP_016869353.1 | |
| CB216817.1 | ||||
| U47742.1 | 17 | 7592 | AAC50662.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831300 | Chr.8:41818817 - 41998976 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map