Assay Details
Target Gene Details
Entrez Gene ID: | 6690 |
Gene Name: | serine peptidase inhibitor, Kazal type 1 |
Gene Aliases: |
PCTT, PSTI, Spink3, TATI, TCP |
Location: |
Chr.5:147824580-147839231 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPINK1 | NM_003122.4 | NP_003113.2 | ||
| AI990990.1 | ||||
| CB159576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1021356 | Chr.5:147724064 - 147930968 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map