Assay Details
Target Gene Details
Entrez Gene ID: | 92610 |
Gene Name: | TRAF interacting protein with forkhead associated domain |
Gene Aliases: |
T2BP, T6BP, TIFAA |
Location: |
Chr.4:112275626-112285903 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIFA | NM_052864.2 | 2 | 633 | NP_443096.1 |
| AB062110.1 | 1 | 351 | BAB86902.1 | |
| AB097011.1 | 2 | 635 | BAC77364.1 | |
| AB097038.1 | 2 | 635 | BAC77391.1 | |
| AK292900.1 | 2 | 630 | ||
| BC008294.1 | 2 | 617 | AAH08294.2 | |
| BC014259.2 | 2 | 613 | AAH14259.1 |
More Information
Set Membership: |
|

Genomic Map