Assay Details
Target Gene Details
Entrez Gene ID: | 1650 |
Gene Name: | dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit |
Gene Aliases: |
AGER1, CDG1R, OKSWcl45, OST, OST48, WBP1 |
Location: |
Chr.1:20651767-20661544 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDOST | NM_005216.4 | NP_005207.2 | ||
| AB062391.1 | BAB93478.1 | |||
| AK296041.1 | ||||
| AK297009.1 | ||||
| AK315633.1 | ||||
| BC002594.2 | AAH02594.1 | |||
| D29643.1 | BAA06126.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545731 | Chr.1:19989106 - 20776306 on Build GRCh38 | Gain |
|
| nsv1003348 | Chr.1:20648970 - 20715062 on Build GRCh38 | Gain |
|
| nsv834369 | Chr.1:20609770 - 20779837 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map