Assay Details
Target Gene Details
Entrez Gene ID: | 26133 |
Gene Name: | transient receptor potential cation channel subfamily C member 4 associated protein |
Gene Aliases: |
C20orf188, PPP1R158, TRRP4AP, TRUSS |
Location: |
Chr.20:35002404-35092837 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPC4AP | NM_015638.2 | 19 | 2940 | NP_056453.1 |
| NM_199368.1 | 19 | 2916 | NP_955400.1 | |
| XM_011528774.1 | 14 | 2253 | XP_011527076.1 | |
| XM_017027800.1 | 18 | 2754 | XP_016883289.1 | |
| XM_017027801.1 | 17 | 2418 | XP_016883290.1 | |
| AF055022.1 | 10 | 1643 | ||
| AK074106.1 | 18 | 4455 | BAB84932.1 | |
| AL096738.1 | 2 | 4171 | ||
| AL117480.1 | 13 | 2196 | CAB55953.1 | |
| BC001323.1 | 8 | 1453 | AAH01323.1 | |
| BC008836.2 | 6 | 1306 | AAH08836.2 | |
| BC013144.1 | 19 | 2909 | AAH13144.1 | |
| BM723903.1 | 1 | 244 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063673 | Chr.20:34977862 - 35009073 on Build GRCh38 | Loss |
|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
| nsv833962 | Chr.20:34927688 - 35113250 on Build GRCh38 | Loss |
|
| nsv3362 | Chr.20:34978502 - 35010725 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map