Assay Details
Target Gene Details
Entrez Gene ID: | 5153 |
Gene Name: | phosphodiesterase 1B |
Gene Aliases: |
HEL-S-79p, PDE1B1, PDES1B |
Location: |
Chr.12:54549393-54579239 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 15 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE1B | NM_000924.3 | NP_000915.1 | ||
| NM_001165975.2 | NP_001159447.1 | |||
| NM_001288768.1 | NP_001275697.1 | |||
| NM_001288769.1 | NP_001275698.1 | |||
| NM_001315534.1 | NP_001302463.1 | |||
| NM_001315535.1 | NP_001302464.1 | |||
| XM_011538456.1 | XP_011536758.1 | |||
| XM_017019432.1 | XP_016874921.1 | |||
| XM_017019433.1 | XP_016874922.1 | |||
| AB209200.1 | 9 | 3451 | BAD92437.1 | |
| AJ401609.1 | CAC82207.1 | |||
| AK126899.1 | ||||
| AK296422.1 | ||||
| AK302899.1 | ||||
| AK302931.1 | ||||
| BC032226.1 | AAH32226.1 | |||
| BX538092.1 | CAD98011.1 | |||
| EU668337.1 | 9 | 3451 | ||
| U40584.1 | AAB50018.1 | |||
| U56976.1 | AAC50769.1 | |||
| U86078.1 | AAC51872.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832421 | Chr.12:54522913 - 54691618 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map