Assay Details
Target Gene Details
Entrez Gene ID: | 79039 |
Gene Name: | DEAD-box helicase 54 |
Gene Aliases: |
DP97 |
Location: |
Chr.12:113157173-113185479 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX54 | NM_001111322.1 | NP_001104792.1 | ||
| NM_024072.3 | NP_076977.3 | |||
| AF478457.1 | AAL85336.1 | |||
| AY148094.1 | AAN59978.1 | |||
| BC005848.2 | AAH05848.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv519540 | Chr.12:113042705 - 113459599 on Build GRCh38 | Gain+Loss |
|
| esv3892265 | Chr.12:113099321 - 113527932 on Build GRCh38 | Gain |
|
| dgv2884n54 | Chr.12:113115603 - 113166364 on Build GRCh38 | Loss |
|
| esv996809 | Chr.12:113160789 - 113174565 on Build GRCh38 | Deletion |
|
| esv3630792 | Chr.12:112988703 - 113838810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map