Assay Details
Target Gene Details
Entrez Gene ID: | 10617 |
Gene Name: | STAM binding protein |
Gene Aliases: |
AMSH, MICCAP |
Location: |
Chr.2:73828911-73873656 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAMBP | NM_006463.4 | 3 | 276 | NP_006454.1 |
| NM_201647.2 | 2 | 167 | NP_964010.1 | |
| NM_213622.2 | 2 | 599 | NP_998787.1 | |
| XM_005264088.3 | 2 | 174 | XP_005264145.1 | |
| XM_011532483.2 | 3 | 271 | XP_011530785.1 | |
| XM_011532484.2 | 3 | 289 | XP_011530786.1 | |
| XM_011532485.2 | 3 | 271 | XP_011530787.1 | |
| XM_017003175.1 | 3 | 222 | XP_016858664.1 | |
| XM_017003176.1 | 2 | 172 | XP_016858665.1 | |
| XM_017003177.1 | 3 | 277 | XP_016858666.1 | |
| XM_017003178.1 | 2 | 625 | XP_016858667.1 | |
| XM_017003179.1 | 3 | 223 | XP_016858668.1 | |
| XM_017003180.1 | 2 | 156 | XP_016858669.1 | |
| XM_017003181.1 | 2 | 614 | XP_016858670.1 | |
| AF052135.1 | 2 | 190 | ||
| BC007682.1 | 1 | 90 | AAH07682.1 | |
| BC065574.1 | 2 | 577 | AAH65574.1 | |
| BC101467.1 | 2 | 165 | AAI01468.1 | |
| BC101469.1 | 2 | 163 | AAI01470.1 | |
| BM478027.1 | 3 | 263 | ||
| BQ929694.1 | 2 | 125 | ||
| CX758449.1 | 2 | 599 | ||
| DB477012.1 | 3 | 276 | ||
| DN919763.1 | 2 | 182 | ||
| DR158757.1 | 3 | 261 | ||
| EU927390.1 | 1 | 69 | ||
| U73522.1 | 1 | 84 | AAD05037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834260 | Chr.2:73678979 - 73849393 on Build GRCh38 | Gain |
|
| esv2759060 | Chr.2:73489905 - 73851465 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map