Assay Details
Target Gene Details
Entrez Gene ID: | 80204 |
Gene Name: | F-box protein 11 |
Gene Aliases: |
FBX11, PRMT9, UBR6, UG063H01, VIT1 |
Location: |
Chr.2:47806920-47905793 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO11 | NM_001190274.1 | NP_001177203.1 | ||
| NM_025133.4 | NP_079409.3 | |||
| XM_005264572.4 | XP_005264629.1 | |||
| XM_005264573.4 | XP_005264630.1 | |||
| XM_017005015.1 | XP_016860504.1 | |||
| XM_017005016.1 | XP_016860505.1 | |||
| XM_017005017.1 | XP_016860506.1 | |||
| AK022735.1 | BAB14214.1 | |||
| AK025477.1 | BAB15143.1 | |||
| AK097734.1 | ||||
| AK223592.1 | BAD97312.1 | |||
| AK292877.1 | ||||
| AK297444.1 | ||||
| AK304140.1 | ||||
| AK315108.1 | ||||
| AY827075.1 | AAV87312.1 | |||
| BC012728.2 | AAH12728.2 | |||
| BC043258.1 | AAH43258.2 | |||
| BC130445.1 | ||||
| BC136480.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs77079615] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map