Assay Details
Target Gene Details
Entrez Gene ID: | 762 |
Gene Name: | carbonic anhydrase 4 |
Gene Aliases: |
CAIV, Car4, RP17 |
Location: |
Chr.17:60149941-60178118 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CA4 | XM_005257639.2 | XP_005257696.1 | ||
| XM_011525183.2 | XP_011523485.1 | |||
| XM_017025012.1 | XP_016880501.1 | |||
| AI990988.1 | ||||
| AK289715.1 | ||||
| AK298710.1 | ||||
| BC057792.1 | AAH57792.1 | |||
| BC069649.1 | AAH69649.1 | |||
| BC074768.2 | AAH74768.1 | |||
| CR541766.1 | CAG46565.1 | |||
| M83670.1 | AAA35630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527094 | Chr.17:60128778 - 60163403 on Build GRCh38 | Loss |
|
| nsv519766 | Chr.17:60148405 - 60163403 on Build GRCh38 | Loss |
|
| nsv9579 | Chr.17:60081156 - 60159528 on Build GRCh38 | Gain+Loss |
|
| nsv833504 | Chr.17:60071000 - 60277790 on Build GRCh38 | Gain |
|
| nsv528050 | Chr.17:60149439 - 60163403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map