Assay Details
Target Gene Details
Entrez Gene ID: | 85329 |
Gene Name: | galectin 12 |
Gene Aliases: |
GAL12, GRIP1 |
Location: |
Chr.11:63506052-63516774 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGALS12 | NM_001142535.1 | NP_001136007.1 | ||
| NM_001142536.1 | NP_001136008.1 | |||
| NM_001142537.1 | NP_001136009.1 | |||
| NM_001142538.1 | NP_001136010.1 | |||
| NM_033101.3 | NP_149092.2 | |||
| XM_017018458.1 | XP_016873947.1 | |||
| AF222694.1 | AAF34676.1 | |||
| AF222695.1 | AAF34677.1 | |||
| AF244974.1 | AAK77328.1 | |||
| AF244975.1 | AAK77329.1 | |||
| AF244976.1 | AAK77330.1 | |||
| AF244977.1 | AAK77331.1 | |||
| AF310686.1 | AAG40863.1 | |||
| AF310687.1 | AAG40864.1 | |||
| AK313851.1 | ||||
| BC028222.1 | AAH28222.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555179 | Chr.11:63465891 - 63552767 on Build GRCh38 | Loss |
|
| nsv509417 | Chr.11:63485643 - 63569743 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map