Assay Details
Target Gene Details
Entrez Gene ID: | 84064 |
Gene Name: | haloacid dehalogenase like hydrolase domain containing 2 |
Gene Aliases: |
3110052N05Rik, HEL-S-301 |
Location: |
Chr.18:47107402-47150520 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDHD2 | NM_001318765.1 | 5 | 679 | NP_001305694.1 |
| NM_032124.4 | 5 | 622 | NP_115500.1 | |
| XM_005258371.4 | 5 | 642 | XP_005258428.1 | |
| XM_006722557.2 | 6 | 714 | XP_006722620.1 | |
| XM_011526227.2 | 6 | 698 | XP_011524529.1 | |
| XM_011526228.2 | 6 | 696 | XP_011524530.1 | |
| XM_017026039.1 | 6 | 735 | XP_016881528.1 | |
| XM_017026040.1 | 6 | 714 | XP_016881529.1 | |
| AK054590.1 | 5 | 659 | BAB70768.1 | |
| AK292098.1 | 5 | 622 | ||
| AL136681.1 | 5 | 613 | CAB66616.1 | |
| BC011894.2 | 5 | 643 | AAH11894.1 | |
| BC033031.1 | 5 | 658 | AAH33031.1 | |
| BC038100.2 | 5 | 622 | AAH38100.1 | |
| CR533487.1 | 4 | 489 | CAG38518.1 | |
| FJ224322.1 | 5 | 613 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3642451 | Chr.18:47089642 - 47219390 on Build GRCh38 | Gain |
|
| nsv576819 | Chr.18:47075878 - 47151256 on Build GRCh38 | Loss |
|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| nsv833640 | Chr.18:47025905 - 47188501 on Build GRCh38 | Loss |
|
| esv3642452 | Chr.18:47093035 - 47161369 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map