Assay Details
Target Gene Details
Entrez Gene ID: | 64641 |
Gene Name: | early B-cell factor 2 |
Gene Aliases: |
COE2, EBF-2, O/E-3, OE-3 |
Location: |
Chr.8:25841730-26045124 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBF2 | NM_022659.3 | 16 | 2039 | NP_073150.2 |
| AK021562.1 | 9 | 1078 | BAB13843.1 | |
| AK304372.1 | 15 | 1687 | ||
| AY700779.1 | AAU10086.1 | |||
| BC069665.1 | 9 | 1056 | AAH69665.1 | |
| BC069726.1 | 9 | 1059 | AAH69726.1 | |
| BC069747.1 | 9 | 1058 | AAH69747.1 | |
| BC069768.1 | 9 | 1059 | AAH69768.1 | |
| BC074794.2 | AAH74794.2 | |||
| BC113478.1 | 8 | 1023 | ||
| BC113504.1 | 8 | 1023 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161746 | Chr.8:25843111 - 25850751 on Build GRCh38 | Deletion |
|
| esv2759607 | Chr.8:25823806 - 25849698 on Build GRCh38 | Loss |
|
| esv32961 | Chr.8:25842136 - 25943866 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map