Assay Details
Target Gene Details
Entrez Gene ID: | 8325 |
Gene Name: | frizzled class receptor 8 |
Gene Aliases: |
FZ-8, hFZ8 |
Location: |
Chr.10:35638249-35641434 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FZD8 | NM_031866.2 | 1 | 2031 | NP_114072.1 |
| AB043703.1 | 1 | 2031 | BAB41064.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv144n67 | Chr.10:35600010 - 35681917 on Build GRCh38 | Gain |
|
| nsv521143 | Chr.10:35624277 - 35647825 on Build GRCh38 | Loss |
|
| esv22469 | Chr.10:35639220 - 35642926 on Build GRCh38 | Loss |
|
| nsv466871 | Chr.10:35600599 - 35673959 on Build GRCh38 | Loss |
|
| nsv825340 | Chr.10:35628179 - 35651436 on Build GRCh38 | Gain |
|
| nsv825341 | Chr.10:35638193 - 35643553 on Build GRCh38 | Gain |
|
| nsv825337 | Chr.10:35583021 - 35685326 on Build GRCh38 | Gain |
|
| dgv1035n54 | Chr.10:35631696 - 35644550 on Build GRCh38 | Loss |
|
| dgv1036n54 | Chr.10:35638239 - 35642852 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map