Assay Details
Target Gene Details
Entrez Gene ID: | 30061 |
Gene Name: | solute carrier family 40 member 1 |
Gene Aliases: |
FPN1, HFE4, IREG1, MST079, MSTP079, MTP1, SLC11A3 |
Location: |
Chr.2:189560590-189580811 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC40A1 | NM_014585.5 | 8 | 1969 | NP_055400.1 |
| XM_005246505.1 | 9 | 2104 | XP_005246562.1 | |
| XM_017003938.1 | 9 | 2218 | XP_016859427.1 | |
| AF147322.1 | 1 | 217 | ||
| AF215636.1 | 8 | 1922 | AAF80986.1 | |
| AF226614.1 | 8 | 1922 | AAF36697.1 | |
| AF231121.1 | 8 | 1922 | AAF44330.1 | |
| AK002038.1 | 3 | 902 | BAA92049.1 | |
| AK223236.1 | 8 | 1944 | BAD96956.1 | |
| AK292146.1 | 8 | 1944 | ||
| AK314827.1 | 8 | 1944 | ||
| AL136944.1 | 8 | 1932 | CAB66878.1 | |
| BC035893.1 | 8 | 1933 | AAH35893.1 | |
| BC037733.1 | 8 | 2048 | AAH37733.1 | |
| BX537503.1 | 1 | 1798 | ||
| CR533564.1 | 8 | 1618 | CAG38595.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954448 | Chr.2:189550075 - 189580174 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map