Assay Details
Target Gene Details
Entrez Gene ID: | 8761 |
Gene Name: | poly(A) binding protein cytoplasmic 4 |
Gene Aliases: |
APP-1, APP1, PABP4, iPABP |
Location: |
Chr.1:39560813-39576849 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PABPC4 | NM_001135653.1 | 1 | 685 | NP_001129125.1 |
| NM_001135654.1 | 1 | 685 | NP_001129126.1 | |
| NM_003819.3 | 1 | 685 | NP_003810.1 | |
| BC065540.1 | 1 | 587 | AAH65540.1 | |
| BC071591.1 | 1 | 623 | AAH71591.1 | |
| BC094755.1 | 1 | 626 | AAH94755.1 | |
| U75686.1 | 1 | 580 | AAB97309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829649 | Chr.1:39505283 - 39638842 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map