Assay Details
Target Gene Details
Entrez Gene ID: | 93664 |
Gene Name: | calcium dependent secretion activator 2 |
Gene Aliases: |
CAPS2 |
Location: |
Chr.7:122318424-122886759 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Gene Details
Entrez Gene ID: | 168433 |
Gene Name: | ring finger protein 133 |
Gene Aliases: |
- |
Location: |
Chr.7:122697712-122699154 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF133 | NM_139175.1 | 1 | 1292 | NP_631914.1 |
| AF447589.1 | 1 | 1292 | AAM22872.1 | |
| AK098524.1 | 1 | 1294 | BAC05321.1 | |
| BC022038.2 | 1 | 1271 | AAH22038.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map