Assay Details
Target Gene Details
Entrez Gene ID: | 9619 |
Gene Name: | ATP binding cassette subfamily G member 1 |
Gene Aliases: |
ABC8, WHITE1 |
Location: |
Chr.21:42199689-42304389 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCG1 | NM_207627.1 | NP_997510.1 | ||
| NM_207628.1 | NP_997511.1 | |||
| AF323660.1 | AAK28838.1 | |||
| AF323661.1 | AAK28839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762113 | Chr.21:42199305 - 42211517 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map