Assay Details
Target Gene Details
Entrez Gene ID: | 16 |
Gene Name: | alanyl-tRNA synthetase |
Gene Aliases: |
CMT2N, EIEE29 |
Location: |
Chr.16:70252295-70289552 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AARS | NM_001605.2 | 21 | 3255 | NP_001596.2 |
| AK222824.1 | 21 | 3252 | BAD96544.1 | |
| AK225116.1 | ||||
| AK299098.1 | ||||
| BC011451.1 | 21 | 3187 | AAH11451.1 | |
| BU178772.1 | 4 | 664 | ||
| D32050.1 | 21 | 3255 | BAA06808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33745 | Chr.16:70082696 - 70285970 on Build GRCh38 | Gain+Loss |
|
| esv2758652 | Chr.16:69821219 - 70279319 on Build GRCh38 | Gain+Loss |
|
| nsv1061286 | Chr.16:70219906 - 70297504 on Build GRCh38 | Gain |
|
| nsv428328 | Chr.16:69952660 - 70279319 on Build GRCh38 | Gain+Loss |
|
| nsv833271 | Chr.16:70151791 - 70319431 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116553521] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map