Assay Details
Target Gene Details
Entrez Gene ID: | 259239 |
Gene Name: | WAP four-disulfide core domain 11 |
Gene Aliases: |
WAP11 |
Location: |
Chr.20:45648563-45670239 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC11 | NM_147197.2 | NP_671730.1 | ||
| AY047609.1 | AAK97771.1 | |||
| BC062670.1 | AAH62670.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586081 | Chr.20:45573240 - 45747557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map