Assay Details
Target Gene Details
Entrez Gene ID: | 118672 |
Gene Name: | phosphoseryl-tRNA kinase |
Gene Aliases: |
C10orf89 |
Location: |
Chr.10:122980040-122997519 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSTK | NM_153336.2 | NP_699167.2 | ||
| XM_017015640.1 | XP_016871129.1 | |||
| XM_017015641.1 | XP_016871130.1 | |||
| AK094271.1 | 1 | 772 | ||
| AK127173.1 | BAC86867.1 | |||
| AL833506.1 | 1 | 293 | ||
| BC035344.1 | AAH35344.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7586 | Chr.10:122982687 - 123016052 on Build GRCh38 | Insertion |
|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map