Assay Details
Target Gene Details
Entrez Gene ID: | 1021 |
Gene Name: | cyclin dependent kinase 6 |
Gene Aliases: |
MCPH12, PLSTIRE |
Location: |
Chr.7:92604921-92836627 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDK6 | NM_001145306.1 | 8 | 8733 | NP_001138778.1 |
| NM_001259.6 | 8 | 8628 | NP_001250.1 | |
| XM_006715835.2 | 8 | 8853 | XP_006715898.1 | |
| AB073601.1 | 1 | 633 | ||
| AK055922.1 | 1 | 1601 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819408 | Chr.7:92607938 - 92608315 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map