Assay Details
Target Gene Details
Entrez Gene ID: | 125111 |
Gene Name: | gap junction protein delta 3 |
Gene Aliases: |
CX31.9, Cx30.2, GJA11, GJC1 |
Location: |
Chr.17:40360652-40364693 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJD3 | NM_152219.3 | NP_689343.3 | ||
| AK125254.1 | BAC86102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833445 | Chr.17:40310222 - 40481717 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map