Assay Details
Target Gene Details
Entrez Gene ID: | 392509 |
Gene Name: | ADP ribosylation factor like GTPase 13A |
Gene Aliases: |
ARL13, dJ341D10.2 |
Location: |
Chr.X:100969011-100990831 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARL13A | NM_001162491.1 | NP_001155963.1 | ||
| NR_103708.1 | ||||
| NR_103709.1 | ||||
| XM_011530950.2 | XP_011529252.1 | |||
| AK301813.1 | ||||
| BC140808.1 | ||||
| BC144603.1 | ||||
| CD300373.1 |
Target Gene Details
Entrez Gene ID: | 79979 |
Gene Name: | tRNA methyltransferase 2 homolog B |
Gene Aliases: |
CXorf34, dJ341D10.3 |
Location: |
Chr.X:100973358-101052118 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT2B |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517937 | Chr.X:100982949 - 101015369 on Build GRCh38 | Gain |
|
| nsv515758 | Chr.X:100987316 - 101038566 on Build GRCh38 | Gain |
|
| nsv7012 | Chr.X:100965059 - 100999697 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map