Assay Details
Target Gene Details
Entrez Gene ID: | 326625 |
Gene Name: | methylmalonic aciduria (cobalamin deficiency) cblB type |
Gene Aliases: |
ATR, CFAP23, cblB, cob |
Location: |
Chr.12:109553715-109573553 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMAB | NM_052845.3 | 9 | 1270 | NP_443077.1 |
| NR_038118.1 | 10 | 1357 | ||
| BC011831.2 | 9 | 1214 | AAH11831.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560118 | Chr.12:109510100 - 109607113 on Build GRCh38 | Loss |
|
| esv3630719 | Chr.12:109485009 - 109700854 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map