Assay Details
Target Gene Details
Entrez Gene ID: | 3508 |
Gene Name: | immunoglobulin mu binding protein 2 |
Gene Aliases: |
CATF1, CMT2S, HCSA, HMN6, SMARD1, SMUBP2, ZFAND7 |
Location: |
Chr.11:68903851-68940602 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGHMBP2 | NM_002180.2 | NP_002171.2 | ||
| XM_005273976.1 | XP_005274033.1 | |||
| XM_017017671.1 | XP_016873160.1 | |||
| DB448398.1 |
Target Gene Details
Entrez Gene ID: | 219927 |
Gene Name: | mitochondrial ribosomal protein L21 |
Gene Aliases: |
L21mt, MRP-L21 |
Location: |
Chr.11:68891276-68903835 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL21 | NM_181514.1 | NP_852615.1 | ||
| NM_181515.1 | NP_852616.1 | |||
| XM_005273823.4 | XP_005273880.1 | |||
| AK096756.1 | ||||
| AK301989.1 | ||||
| BC055088.1 | AAH55088.1 | |||
| BM916618.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509423 | Chr.11:68873995 - 68970585 on Build GRCh38 | Insertion |
|
| nsv428261 | Chr.11:68754616 - 68937835 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map