Assay Details
Target Gene Details
Entrez Gene ID: | 2548 |
Gene Name: | glucosidase alpha, acid |
Gene Aliases: |
LYAG |
Location: |
Chr.17:80101526-80119882 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GAA | NM_000152.4 | 1 | 201 | NP_000143.2 |
| NM_001079803.2 | NP_001073271.1 | |||
| NM_001079804.2 | NP_001073272.1 | |||
| NR_134848.1 | ||||
| XM_005257193.2 | XP_005257250.1 | |||
| XM_005257194.4 | XP_005257251.1 | |||
| BC040431.1 | AAH40431.1 | |||
| DC392486.1 | ||||
| HY015584.1 | 1 | 95 | ||
| HY047261.1 | 1 | 105 | ||
| HY101855.1 | ||||
| Y00839.1 | CAA68763.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066606 | Chr.17:79955666 - 80224006 on Build GRCh38 | Gain |
|
| nsv469591 | Chr.17:80085322 - 80293486 on Build GRCh38 | Loss |
|
| nsv833561 | Chr.17:79977946 - 80167051 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map